Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.04971.1000 1.04971.9050
EM1.04971.1000CS 5021 USD
EM1.04971.1000 EM1.04971.9050
Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), Supelco®
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
Density: 1.85 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
UN: 3077
ADR: 9,III
Merck Index: 13,07712

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR