Potassium cyanide, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.04965.1000
EM1.04965.1000CS 879 USD
EM1.04965.1000
Potassium cyanide, Supelco®
Potassium cyanide
Formula: KCN
MW: 65.12 g/mol
Boiling Pt: 1625 °C (1013 hPa)
Melting Pt: 634 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1,I
Merck Index: 13,07709

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR