Potassium chromate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.04952.0250 1.04952.1000
EM1.04952.0250CS 1197 USD
EM1.04952.0250 EM1.04952.1000
Potassium chromate, EMSURE® for analysis, Supelco®
Potassium chromate
Formula: K₂CrO₄
MW: 194.19 g/mol
Boiling Pt: 1000
Melting Pt: 985
Density: 2.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3288
ADR: 6.1 II
Merck Index: 12,07784

Order Now

Specification Test Results

Solubility 637 g/l (20 °C)
Melting Point 985 °C
Molar Mass 194.19 g/mol
Bulk Density 1400 kg/m3
Boiling Point 1000 °C
Density 2.73 g/cm3 (18 °C)
pH 9.0 - 9.8 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR