Potassium bromide, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.04905.0500
EM1.04905.0500CS 871 USD
EM1.04905.0500
Potassium bromide, EMSURE® for analysis, Supelco®
Potassium bromide

In North America 1.04905.0500 replaces but has a far superior specification to PX1380-1

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

Solubility 650 g/l (20 °C)
Melting Point 730 °C
Molar Mass 119.00 g/mol
Bulk Density 900 - 1000 kg/m3
Boiling Point 1435 °C
Vapor Pressure 1.3 hPa (795 °C)
Density 2.75 g/cm3 (20 °C)
pH 5.5 - 8.5 (50 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR