Potassium dihydrogen phosphate 1/15 0.0667 M in aqueous solution buffer stock solution, Supelco®

Supplier: MilliporeSigma

1.04875.1000
EM1.04875.1000CS 511 USD
EM1.04875.1000
Potassium dihydrogen phosphate 1/15 0.0667 M in aqueous solution buffer stock solution, Supelco®
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4

Order Now

Specification Test Results

Density 1.001 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR