Potassium dichromate, EMSURE® ACS, ISO for analysis (max. 0.01 ppm Hg), Supelco®

Supplier: MilliporeSigma
Danger

1.04865.0500
EM1.04865.0500CS 2418 USD
EM1.04865.0500
Potassium dichromate, EMSURE® ACS, ISO for analysis (max. 0.01 ppm Hg), Supelco®
Potassium dichromate
Formula: K₂Cr₂O₇
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 398 °C
Density: 2.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3086
ADR: 6.1 (5.1),II
REACH: 01-2119454792-32
Merck Index: 12,07790

Order Now

Specification Test Results

Solubility 130 g/l (20 °C)
Melting Point 398 °C
Molar Mass 294.19 g/mol
Bulk Density 1250 kg/m3
Boiling Point 500 °C (1013 hPa)
Density 2.69 g/cm3 (20 °C)
pH 3.6 (100 g/l, H2O)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR