Potassium dichromate, EMSURE® ACS, Reag. Ph. Eur., ISO for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.04864.0500
1.04864.1000
EM1.04864.0500CS
1613
USD
EM1.04864.0500
EM1.04864.1000
EM1.04864.5000
Potassium dichromate, EMSURE® ACS, Reag. Ph. Eur., ISO for analysis, Supelco®
Potassium dichromate
1.04864.0500 replaces and is the same as PX1445-1. Part # Change only, Same Pack Size, Same Specifications.
1.04864.0500 will be offered as an alternative to PX1445-11. Different Pack Size, Same Specifications.
1.04864.1000 replaces and is the same as PX1443-6. Part # Change only, Same Pack Size, Same Specifications.
1.04864.1000 will be offered as an alternative to PX1445-2. Part # Change only, Same Pack Size, Same Specifications.
Formula:
K₂Cr₂O₇ MW: 294.18 g/mol Boiling Pt: 500 °C (1013 hPa) Melting Pt: 398 °C Density: 2.67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011367 CAS Number: 7778-50-9 EINECS: 231-906-6 UN: 3086 ADR: 6.1 (5.1),II REACH: 01-2119454792-32 Merck Index: 12,07790 |
Specification Test Results
Solubility | 130 g/l (20 °C) |
Melting Point | 398 °C |
Molar Mass | 294.19 g/mol |
Bulk Density | 1250 kg/m3 |
Boiling Point |
|
Density | 2.69 g/cm3 (20 °C) |
pH | 3.6 (100 g/l, H2O) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...