Potassium dichromate, EMSURE® ACS, Reag. Ph. Eur., ISO for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.04864.0500 1.04864.1000
EM1.04864.0500CS 1613 USD
EM1.04864.0500 EM1.04864.1000 EM1.04864.5000
Potassium dichromate, EMSURE® ACS, Reag. Ph. Eur., ISO for analysis, Supelco®
Potassium dichromate

1.04864.0500 replaces and is the same as PX1445-1. Part # Change only, Same Pack Size, Same Specifications.
1.04864.0500 will be offered as an alternative to PX1445-11. Different Pack Size, Same Specifications.
1.04864.1000 replaces and is the same as PX1443-6. Part # Change only, Same Pack Size, Same Specifications.
1.04864.1000 will be offered as an alternative to PX1445-2. Part # Change only, Same Pack Size, Same Specifications.

Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 398 °C
Density: 2.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3086
ADR: 6.1 (5.1),II
REACH: 01-2119454792-32
Merck Index: 12,07790

Order Now

Specification Test Results

Solubility 130 g/l (20 °C)
Melting Point 398 °C
Molar Mass 294.19 g/mol
Bulk Density 1250 kg/m3
Boiling Point 500 °C (1013 hPa)
Density 2.69 g/cm3 (20 °C)
pH 3.6 (100 g/l, H2O)
Hazard Class 6.1
Hazard Class Description Toxic materials

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR