Potassium chloride 2.97-3.03 M saturated with silver chloride aqueous solution, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Potassiumchloride

1.04818.0100
EM1.04818.0100CS 3200 USD
EM1.04818.0100
Potassium chloride 2.97-3.03 M saturated with silver chloride aqueous solution, Supelco®
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 100 °C (1013 hPa)
Density 1.13 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR