Iodine, EMSURE® for analysis, sublimated, Supelco®

Supplier: MilliporeSigma
Danger

1.04761.0500
EM1.04761.0100CS 9019.01 USD
EM1.04761.0100 EM1.04761.0500
Iodine, EMSURE® for analysis, sublimated, Supelco®
Iodine
Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8 (6.1),III

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point 114 °C
Molar Mass 253.8 g/mol
Bulk Density 2100 kg/m3
Boiling Point 185 °C (1013 hPa)
Vapor Pressure 0.41 hPa (25 °C)
Density 4.93 g/cm3 (20 °C)
pH 5.4 (H2O) (saturated solution)
Viscosity Kinematic 0.57 mm2/s (116 °C) liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR