UV-VIS Standard 1a: Potassium Dichromate Solution (600 mg/L) for the Absorbance at 430 nm According to Ph Eur Certipur®
Supplier: MilliporeSigma
1.04660.0001
EM1.04660.0001CS
23134
USD
EM1.04660.0001
UV-VIS Standard 1a: Potassium Dichromate Solution (600 mg/L) for the Absorbance at 430 nm According to Ph Eur Certipur®
Standards
Color Reference and Spectroscopy Standards
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)