Hydroxylammonium chloride ACS, ISO, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

M-Clarity™ quality level = MQ100

1.04619.0250
EM1.04619.0250EA 455 USD
EM1.04619.0250
Hydroxylammonium chloride ACS, ISO, Supelco®
Hydroxylammonium chloride

Hydroxylammonium chloride for analysis (≤ 0.000001% Hg) ACS,ISO.

Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Specification Test Results

Assay (manganometric) ≥ 99.0 %
Clarity of the solution in Ethanol passes test
pH-value (5 %; water) 2.5 - 3.5
Titrable free acid ≤ 0.25 meq/g
Sulfate (SO₄) ≤ 0.002 %
Heavy metals (ACS) ≤ 0.0005 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.0005 %
Hg (Mercury) ≤ 0.000001 %
NH₄ (Ammonium) passes test
Pb (Lead) ≤ 0.0005 %
Total sulfur (as sulfate) ≤ 0.005 %
Residue on ignition (as sulfate) ≤ 0.01 %
Corresponds to ACS,ISO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR