Hydroxylammonium sulfate ≥99%, GR analytical reagent, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Bis(hydroxylammonium) sulphate, Hydroxylamine sulfate (2:1), Hydroxylamine sulphate, Hydroxylamine sulphate (2:1), Bis(hydroxylammonium) sulfate, Hydroxylamine sulfate
M-Clarity™ quality level = MQ100
1.04617.0500
EM1.04617.0500CS
1943
USD
EM1.04617.0500
Hydroxylammonium sulfate ≥99%, GR analytical reagent, Supelco®
Hydroxylammonium sulphate
Formula:
H₈N₂O₆S MW: 164.14 g/mol Melting Pt: 170 °C Density: 1.1 g/cm³ (20 °C) |
MDL Number:
MFCD00044869 CAS Number: 10039-54-0 EINECS: 233-118-8 UN: 2865 ADR: 8,III Merck Index: 13,04853 |
Specification Test Results
Assay | ≥ 99 % |
Chloride (Cl) | ≤ 0.001 % |
As (Arsenic) | ≤ 0.00005 % |
Cu (Copper) | ≤ 0.0005 % |
Fe (Iron) | ≤ 0.0005 % |
Hg (Mercury) | ≤ 0.000001 % |
Pb (Lead) | ≤ 0.0005 % |
Sulfated ash (residue on ignition) | ≤ 0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...