Hydroxylammonium chloride Reag. Ph. Eur., ACS, ISO, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

M-Clarity™ quality level = MQ100

1.04616.0250 1.04616.1000
EM1.04616.0250EA 111 USD
EM1.04616.0250 EM1.04616.1000
Hydroxylammonium chloride Reag. Ph. Eur., ACS, ISO, GR for analysis, Supelco®
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306
Melting Pt: 151
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 III
Merck Index: 13,04853

Order Now

Specification Test Results

Assay (manganometric) ≥ 99.0 %
Clarity of the solution in Ethanol passes test
pH-value (5 %; water) 2.5 - 3.5
Titrable free acid ≤ 0.25 meq/g
Sulfate (SO₄) ≤ 0.002 %
Heavy metals (ACS) ≤ 0.0005 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.0005 %
NH₄ (Ammonium) passes test
Pb (Lead) ≤ 0.0005 %
Total sulfur (as sulfate) ≤ 0.005 %
Residue on ignition (as sulfate) ≤ 0.01 %
Corresponds to ACS, ISO, Reag. Ph Eur


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR