Calcon® (mordant black 17) indicator for metal titration (C.I.15705), Supelco®

Supplier: MilliporeSigma

1.04594.0050
EM1.04594.0050CS 1207 USD
EM1.04594.0050
Calcon® (mordant black 17) indicator for metal titration (C.I.15705), Supelco®
Calcon® (Mordant Black 17)
Formula: C₂₀H₁₃N₂NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00004069
CAS Number: 2538-85-4
EINECS: 219-810-2

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Molar Mass 416.39 g/mol
Bulk Density 530 kg/m3
pH 9.4 (10 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR