Mercury(II) thiocyanate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.04484.0025
1.04484.0100
EM1.04484.0025CS
23459
USD
EM1.04484.0025
EM1.04484.0100
Mercury(II) thiocyanate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) thiocyanate
Formula:
C₂HgN₂S₂ Melting Pt: –38.9 °C Density: 4 g/cm³ (20 °C) Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011039 CAS Number: 592-85-8 EINECS: 209-773-0 UN: 1646 ADR: 6.1,II Merck Index: 13,05917 |
Specification Test Results
Assay (complexometric) | ≥ 99.0 % |
Fe (Iron) | ≤ 0.002 % |
Substances not reducing by formic acid (residue on ignition) | ≤ 0.05 % |
Corresponds to Reag. Ph Eur |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...