Mercury(II) thiocyanate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.04484.0025 1.04484.0100
EM1.04484.0025CS 23459 USD
EM1.04484.0025 EM1.04484.0100
Mercury(II) thiocyanate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) thiocyanate
Formula: C₂HgN₂S₂
Melting Pt: –38.9 °C
Density: 4 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00011039
CAS Number: 592-85-8
EINECS: 209-773-0
UN: 1646
ADR: 6.1,II
Merck Index: 13,05917

Order Now

Specification Test Results

Assay (complexometric) ≥ 99.0 %
Fe (Iron) ≤ 0.002 %
Substances not reducing by formic acid (residue on ignition) ≤ 0.05 %
Corresponds to Reag. Ph Eur


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR