Mercury(II) acetate ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt
1.04410.0050
1.04410.0250
EM1.04410.0050CS
16441
USD
EM1.04410.0050
EM1.04410.0250
Mercury(II) acetate ACS, Reag. Ph. Eur. for analysis, Supelco®
Mercury (II) acetate
Formula:
(H₃CCOO)₂Hg Melting Pt: 178…180 °C Density: 3.27 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012165 CAS Number: 1600-27-7 EINECS: 216-491-1 UN: 1629 ADR: 6.1,II Merck Index: 13,05898 |
Specification Test Results
Solubility | 400 g/l (20 °C) |
Melting Point | 178 - 180 °C (decomposition) |
Molar Mass | 318.68 g/mol |
Bulk Density | 1000 kg/m3 |
Density | 3.27 g/cm3 (20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...