n-Hexane, LiChrosolv® for liquid chromatography, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.04391.1000 1.04391.2500 1.04391.5000 1.04391.4000
EM1.04391.1000CS 849 USD
EM1.04391.1000 EM1.04391.2500 EM1.04391.5000 EM1.04391.4000
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR