n-Heptane ≥99% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: high purity solvents

M-Clarity™ quality level = MQ200

1.04365.1000 1.04365.2500 1.04365.2511 1.04365.6025 1.04365.6190 1.04365.9011
EM1.04365.1000CS 668 USD
EM1.04365.1000 EM1.04365.2500 EM1.04365.2511 EM1.04365.6025 EM1.04365.6190 EM1.04365.9011
n-Heptane ≥99% (by GC), EMPLURA®, Supelco®
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 (lit.)
Melting Pt: -90.6 (lit.)
Density: 0.684 g/ml (25°C) (lit.)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR