Glycerine ≥85%, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma

1.04094.2500
EM1.04094.2500CS 2045 USD
EM1.04094.2500
Glycerine ≥85%, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -10 °C
Boiling Point 130 °C (1013 hPa)
Vapor Pressure 8 hPa (20 °C)
Flash Point 199 °C
Density 1.23 g/cm3 (20 °C)
pH 5 (100 g/l, H2O, 20 °C)
Ignition Point 429 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR