M-GLAS™ Liquid Cover Glass, MilliporeSigma
Supplier: MilliporeSigma
1.03973.0001
EM1.03973.0001EA 145 USD
EM1.03973.0001
M-GLAS™ Liquid Cover Glass, MilliporeSigma
Stains Microscopy and staining reagents
M-GLAS® should be used instead of a cover glass for the homogeneous mounting of cytological specimens
Place a few drops of the mounting medium on the specimen and spread evenly. When the solvent has evaporated, a firm protective surface coating will evolve. (Refractive index (20°C): 1.490 - 1.500). It is registered as IVD and CE product.