Iron(II) sulfide, sticks, fused, Supelco®

Supplier: MilliporeSigma

Synonyms: Ferrous sulphide, Ferrous sulfide

M-Clarity™ quality level = MQ100

1.03956.1000 1.03956.9025
EM1.03956.1000CS 977 USD
EM1.03956.1000 EM1.03956.9025
Iron(II) sulfide, sticks, fused, Supelco®
Iron (II) sulphide
Formula: FeS
Melting Pt: 1195 °C
Density: 4.84 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
UN: 3077
ADR: 9,III
Merck Index: 12,04106

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR