Silica gel sachet: 15 x 14 cm desiccant, with moisture indicator (orange gel), Supelco®

Supplier: MilliporeSigma

1.03805.0001
EM1.03805.0001CS 1308 USD
EM1.03805.0001
Silica gel sachet: 15 x 14 cm desiccant, with moisture indicator (orange gel), Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Specification Test Results

Solubility 1 g/l
Melting Point 1000 °C
Water Absorption 250 g/kg (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR