Ammonium iron (II) sulfate hexahydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate
M-Clarity™ quality level = MQ300
1.03792.0500
1.03792.1000
1.03792.5000
EM1.03792.0500CS
3370
USD
EM1.03792.0500
EM1.03792.1000
EM1.03792.5000
Ammonium iron (II) sulfate hexahydrate, EMSURE® for analysis, Supelco®
Ammonium iron (II) sulphate hexahydrate
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O Melting Pt: 100 °C Density: 1.864 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 EINECS: 233-151-8 |
Specification Test Results
Assay (manganometric) | 99.0 - 101.5 % |
pH-value (5 %; water) | 3.0 - 5.0 |
Chloride (Cl) | ≤ 0.001 % |
Phosphate (PO₄) | ≤ 0.002 % |
Iron(III)-salts (Fe³⁺) | ≤ 0.02 % |
Ca (Calcium) | ≤ 0.002 % |
Cu (Copper) | ≤ 0.002 % |
K (Potassium) | ≤ 0.01 % |
Mg (Magnesium) | ≤ 0.01 % |
Mn (Manganese) | ≤ 0.05 % |
Na (Sodium) | ≤ 0.01 % |
Pb (Lead) | ≤ 0.001 % |
Zn (Zinc) | ≤ 0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...