Acetonitrile

Supplier: MilliporeSigma

1.03725.1002 1.03725.2002
EM1.03725.1002EA 347 USD
EM1.03725.1002 EM1.03725.2002
Acetonitrile
Acetonitrile
Formula: H₃CCN
MDL Number: MFCD00001878
CAS Number: 75-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR