n-Heptane ≥99.3% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.03654.2500
EM1.03654.2500EA 397 USD
EM1.03654.2500
n-Heptane ≥99.3% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
n-Heptane

LiChrosolv® hypergrade is accurately tested for LC-MS suitability, and meet all the requirements of modern LC-MS ionization methods (ESI/APCI - positive and negative mode).


Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionization efficiency. The packaging material has been improved to meet LC-MS quality requirements perfectly.

Formula: H₃C(CH₂)₅CH₃
Boiling Pt: 97...98 °C (1013 hPa)
Melting Pt: –90.5 °C
Density: 0.68 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥99.3%
Identity (IR) Conforms
Evaporation Residue ≤2.0 mg/l
Water ≤0.005%
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g
Al (Aluminium) ≤ 10 ppb
Ca (Calcium) ≤10 ppb
Fe (Iron) ≤10 ppb
K (Potassium) ≤5 ppb
Mg (Magnesium) ≤10 ppb
Na (Sodium) ≤10 ppb
Transmission (at 210 nm) ≥50%
Transmission (at 220 nm) ≥80%
Transmission (from 245 nm) ≥98%
Suitable for LC-MS; Intensity of Background Mass Peak Based on Reserpine (APCI/ESI Positive) ≤2 ppb
Suitable for LC-MS; Intensity of Background Mass Peak Based on Reserpine (APCI/ESI Negative) ≤20 ppb
Filtered by 0.2 μm Filter


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR