Deuterium oxide ≥99.95% (Isotopic), MagniSolv™ for NMR spectroscopy pH 7 (H₂O, 20 °C), Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.03428.0100
EM1.03428.0005EA 100 USD
EM1.03428.0005 EM1.03428.0009 EM1.03428.0010 EM1.03428.0100
Deuterium oxide ≥99.95% (Isotopic), MagniSolv™ for NMR spectroscopy pH 7 (H₂O, 20 °C), Sigma-Aldrich®
Deuterium oxide

MagniSolv™ deuterated solvents provide: • Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment. • Reliable deuteration degrees. • Low residual water content. • Determination of water content in two ways.

Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Deuteration degree ≥ 99.95 %
Performance test (NMR-spectrum) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR