Parteck® SI 400 (Sorbitol), EMPROVE® ESSENTIAL Ph Eur,BP,NF,FCC,JSFA,E 420

Supplier: MilliporeSigma

1.03140.2500 1.03140.9053
EM1.03140.2500EA 1194.25 USD
EM1.03140.2500 EM1.03140.9053
Parteck® SI 400 (Sorbitol), EMPROVE® ESSENTIAL Ph Eur,BP,NF,FCC,JSFA,E 420
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MDL Number: MFCD00004708
CAS Number: 50-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR