1,4-Dioxane ≥99% (by GC), EMPLURA®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
M-Clarity™ quality level = MQ200
1.03115.1000
1.03115.2500
1.03115.6025
1.03115.9191
EM1.03115.1000CS
1107
USD
EM1.03115.1000
EM1.03115.2500
EM1.03115.6025
EM1.03115.9191
1,4-Dioxane ≥99% (by GC), EMPLURA®, Supelco®
1,4-Dioxane
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 101.1 Melting Pt: 11.8 Density: 1.03 g/cm3 (20 °C) Flash Pt: 12 Storage Temperature: Ambient |
MDL Number:
MFCD00006571 CAS Number: 123-91-1 EINECS: 204-661-8 UN: 1165 ADR: 3 II Merck Index: 13,03330 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...