N,N-Dimethyl-1,4-phenylenediammonium dichloride, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.03067.0025
EM1.03067.0025CS 4933 USD
EM1.03067.0025
N,N-Dimethyl-1,4-phenylenediammonium dichloride, GR for analysis, Supelco®
N,N-Dimethyl-1,4-phenylenediammonium dichloride
Formula: (CH₃)₂NC₆H₄NH₂·2HCl
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00012991
CAS Number: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II
Merck Index: 13,03280

Order Now

Specification Test Results

Solubility 2000 g/l (20 °C)
Melting Point 208 - 212 °C (decomposition)
Molar Mass 209.11 g/mol
Bulk Density 500 kg/m3
pH 1.6 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR