5-(4-(Dimethylamino)benzylidene)rhodanine, GR for analysis, reagent for silver, Supelco®

Supplier: MilliporeSigma

Synonyms: 4-Dimethylaminobenzalrhodanine, 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinone, p-Dimethylaminobenzalrhodanine, 5-(4-(Dimethylamino)benzylidene)-2-mercaptothiazol-4(5H)-one, p-(Dimethylamino)benzal-5-rhodanine, (5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one

1.03059.0005
EM1.03059.0005CS 3258 USD
EM1.03059.0005
5-(4-(Dimethylamino)benzylidene)rhodanine, GR for analysis, reagent for silver, Supelco®
5-(4-(Dimethylamino)benzylidene)rhodanine
Formula: C₁₂H₁₂N₂OS₂
MW: 264.37 g/mol
MDL Number: MFCD00064857
CAS Number: 536-17-4
EINECS: 208-625-2

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 275 - 280 °C decomposes
Molar Mass 264.37 g/mol
Bulk Density 225 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR