Neocuproine, GR for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: 2,9-Dimethyl-1,10-phenanthroline

1.02968.0005
EM1.02968.0005CS 17895 USD
EM1.02968.0005
Neocuproine, GR for analysis, Supelco®
Neocuproine
Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00004973
CAS Number: 484-11-7
EINECS: 207-601-9
Merck Index: 13,06477

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 159 - 164 °C
Molar Mass 208.26 g/mol
Bulk Density 280 kg/m3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR