Dimethyl sulfoxide ≥99.9% (by GC), EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma

1.02952.1000 1.02952.1011 1.02952.2500 1.02952.2511 1.02952.9025
EM1.02952.1000CS 1410 USD
EM1.02952.1000 EM1.02952.1011 EM1.02952.2500 EM1.02952.2511 EM1.02952.9025
Dimethyl sulfoxide ≥99.9% (by GC), EMSURE® ACS for analysis, Supelco®
Dimethyl sulphoxide
Formula: (CH₃)₂SO
Boiling Pt: 189 °C (lit.)
Melting Pt: 16…19 °C (lit.)
Density: 1.10 g/ml (lit.)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Melting Point 18.4 °C
Molar Mass 78.13 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.48 (20 °C)
Explosion Limit 1.8 - 63.0 %(V)
Density 1.10 g/cm3 (20 °C)
Ignition Point 300 - 302 °C
Saturation concentration 8.0 g/m3 (20 °C)
Viscosity kinematic 2.14 mm2/s


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR