N,N-Dimethylformamide, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

1.02937.2500
EM1.02937.0500CS 1649 USD
EM1.02937.0500 EM1.02937.2500
N,N-Dimethylformamide, Uvasol® for spectroscopy, Supelco®
N,N-Dimethylformamide

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -61 °C
Molar Mass 73.09 g/mol
Boiling Point 153 °C (1013 hPa)
Vapor Pressure 3.77 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.43
Explosion Limit 2.2 - 16 %(V)
Density 0.94 g/cm3 (20 °C)
pH 7 (200 g/l, H2O, 20 °C)
Ignition Point 410 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR