Acetonitrile-[D₃] ≥99% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.02904.0010
EM1.02904.0010EA 143 USD
EM1.02904.0010
Acetonitrile-[D₃] ≥99% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Acetonitrile-[D3]

MagniSolv™ deuterated solvents provide:
• Reliable results in the NMR-spectra by excellent chemical purity and highest isotopic enrichment.
• Reliable deuteration degrees.
• Low residual water content.
• Determination of water content in two ways.

Formula: D₃CCN
Boiling Pt: 80.7 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR