Cyclohexanone ≥99% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Pimelic Ketone

M-Clarity™ quality level = MQ200

1.02888.1000 1.02888.2500 1.02888.6010 1.02888.6025 1.02888.9191
EM1.02888.1000CS 488 USD
EM1.02888.1000 EM1.02888.2500 EM1.02888.6010 EM1.02888.6025 EM1.02888.9191
Cyclohexanone ≥99% (by GC), EMPLURA®, Supelco®
Cyclohexanone
Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155.6
Melting Pt: -31
Density: 0.95 g/cm3 (20 °C)
Flash Pt: 43
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3 III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR