Copper(II) sulfate 0.1 mol/l, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma

Synonyms: Cupric sulphate, Cupric sulfate

M-Clarity™ quality level = MQ100

1.02784.1000
EM1.02784.1000CS 356 USD
EM1.02784.1000
Copper(II) sulfate 0.1 mol/l, Titripur® volumetric solution, Supelco®
Copper (II) sulphate
Formula: CuSO₄
MW: 159.61 g/mol
Density: 1.02 g/m3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C. Miscellaneous Hazardous Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR