Copper(II) oxide, powder, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.02766.0100 1.02766.0500
EM1.02766.0100CS 61978 USD
EM1.02766.0100 EM1.02766.0500
Copper(II) oxide, powder, EMSURE® for analysis, Supelco®
Copper (II) oxide
Formula: CuO
MDL Number: MFCD00010979
CAS Number: 1317-38-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1326 °C (decomposition)
Molar Mass 79.55 g/mol
Bulk Density 500 kg/m3
Density 6.48 g/cm3 (25 °C)
pH 7 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR