Copper(II) oxide, powder, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.02766.0100 1.02766.0500
EM1.02766.0100CS 61978 USD
EM1.02766.0100 EM1.02766.0500
Copper(II) oxide, powder, EMSURE® for analysis, Supelco®
Formula: CuO
MDL Number: MFCD00010979
CAS Number: 1317-38-0
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR