Copper(II) oxide, fine wire ∼ 0.65 x 3 mm for elementary analysis (surface: CuO, core: Cu₂O), Supelco®

Supplier: MilliporeSigma
Warning

1.02765.0250 1.02765.1000
EM1.02765.0250CS 4701 USD
EM1.02765.0250 EM1.02765.1000
Copper(II) oxide, fine wire ∼ 0.65 x 3 mm for elementary analysis (surface: CuO, core: Cu₂O), Supelco®
Copper (II) oxide
Formula: CuO
Melting Pt: 1300 °C
Density: 6.2 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR