Copper(II) oxide, granular for elementary analysis pH 7 (50 g/l, H₂O, 20 °C) (slurry), Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

1.02764.0250
EM1.02764.0250CS 83137 USD
EM1.02764.0250
Copper(II) oxide, granular for elementary analysis pH 7 (50 g/l, H₂O, 20 °C) (slurry), Supelco®
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326 (dec.)
Density: 500 kg/m3
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674

Order Now

Specification Test Results

Assay (complexometric) ≥ 99
Chloride (Cl) ≤ 0.005 %
Total sulfur (as SO₄) ≤ 0.015 %
Total nitrogen (N) ≤ 0.002 %
C (Carbon) ≤ 0.01 %
Bulk density 240 - 270 g/100 ml
Particle size (0.71-2 mm) ≥ 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR