Copper(II) oxide, wire for micro analysis, Supelco®

Supplier: MilliporeSigma

1.02762.5000
EM1.02762.5000CS 23603 USD
EM1.02762.5000
Copper(II) oxide, wire for micro analysis, Supelco®
Copper (II) oxide
Formula: CuO
MDL Number: MFCD00010979
CAS Number: 1317-38-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1300 °C
Molar Mass 79.55 g/mol
Bulk Density 2200 kg/m3
Density 6.2 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR