Copper(II) oxide, powder, EMSURE®, Extra Pure for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.02761.0500
1.02761.9025
EM1.02761.0500CS
4981
USD
EM1.02761.0500
EM1.02761.9025
Copper(II) oxide, powder, EMSURE®, Extra Pure for analysis, Supelco®
Copper (II) oxide
Formula:
CuO Melting Pt: 1326 °C (dec.) Density: 6.48 g/cm3 at 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 1326 °C (decomposition) |
Molar Mass | 79.55 g/mol |
Bulk Density | 500 kg/m3 |
Density | 6.48 g/cm3 (25 °C) |
pH | 7 (50 g/l, H2O, 20 °C) (slurry) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...