Ammonium nitrate solution 1 mol/L DIN 19730 for spectrophotometry, Supelco®

Supplier: MilliporeSigma

1.02596.2500
EM1.02596.2500CS 2398 USD
EM1.02596.2500
Ammonium nitrate solution 1 mol/L DIN 19730 for spectrophotometry, Supelco®
Ammonium nitrate
Formula: NH₄NO₃
MDL Number: MFCD00011425
CAS Number: 6484-52-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 100 °C (1013 hPa)
Density 1.03 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR