Spectroquant® Test Kits, Surfactants (Cell Test), MilliporeSigma
Supplier: MilliporeSigma
1.02552.0001
EM1.02552.0001EA 228 USD
EM1.02552.0001
Spectroquant® Test Kits, Surfactants (Cell Test), MilliporeSigma
Environmental Test Kits
Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.