Chloroform-[D₁] ≥99.96% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.02446.0009 1.02446.0025 1.02446.0100
EM1.02446.0009EA 100 USD
EM1.02446.0009 EM1.02446.0010 EM1.02446.0025 EM1.02446.0100
Chloroform-[D₁] ≥99.96% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Chloroform-[D1]

MagniSolv™ deuterated solvents provide reliable results in the NMR-spectra with extremely low residual water, excellent chemical purity and the highest isotopic enrichment.

Formula: CDCl₃
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64.1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR