Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®

Supplier: MilliporeSigma
Warning

1.02409.0100
EM1.02409.0100CS 9745 USD
EM1.02409.0100
Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9 III
Merck Index: 12,10255

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 419 °C
Molar Mass 65.39 g/mol
Boiling Point 906 °C (1013 hPa)
Density 7.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR