Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Certipur® Reag. USP, Reag. Ph. Eur. volumetric standard, secondary reference material for acidimetry, traceable to NIST Standard Reference Material (SRM), Supelco®

Supplier: MilliporeSigma

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

1.02408.0083
EM1.02408.0083EA 257 USD
EM1.02408.0083
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Certipur® Reag. USP, Reag. Ph. Eur. volumetric standard, secondary reference material for acidimetry, traceable to NIST Standard Reference Material (SRM), Supelco®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

  • 10.2 - 10.6 (6 g/l, H₂O, 20 °C)


Merck have introduced into the range of Certipur® standards some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 219…220 °C (13.3 hPa)
Melting Pt: 171…172 °C
Density: 1.35 g/cm3 (23 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR