Sodium carbonate, Certipur® volumetric standard, secondary reference material for acidimetry, traceable to NIST Standard Reference Material (SRM), Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

1.02405.0080
EM1.02405.0080CS 7279 USD
EM1.02405.0080
Sodium carbonate, Certipur® volumetric standard, secondary reference material for acidimetry, traceable to NIST Standard Reference Material (SRM), Supelco®
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 220 g/l (20 °C)
Melting Point 854 °C
Molar Mass 105.99 g/mol
Bulk Density 1100 kg/m3
Boiling Point 1600 °C (decomposition)
Density 2.53 g/cm3 (20 °C)
pH 11.5 (50 g/l, H2O, 25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR